SP Wilmad-LabGlass Calibration/Measuring Flask Set Supplements, TC Metric System, SP Industries
Supplier: Avantor
Certificates
About this item
These borosilicate glass graduated neck flasks come in sizes to supplement our official metric series, LG-4317
Additional sizes can be supplied on request. Calibrated "to contain" at 68°F/20°C.
Customers Who Bought This Also Bought