About this item
Econo™ Sterile Lorna (Edna) Towel Clamps are a non-perforating clamp used to secure instruments, surgical materials such as tubing, and towels to the drape surrounding a surgical site.
- This clamp should never be used on skin
- Ready-to-use, sterile, disposable, floor-grade instruments
The jaws and tips are curved while the shanks and handle are flat so they can rest on the patient. The teeth on the tips of this towel clamp are blunted so they grip tightly but do not penetrate the surgical drape. They also have ratcheted finger ring handles so a firm hold can be secured. The Lorna towel clamp is also commonly referred to as the Edna towel clamp.
To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Specifications
- Catalog No:
- 89023-808
- 89023-806
- Disposable / reusable:
- Disposable
- Disposable
- Latex/Latex-free:
- Latex-free
- Latex-free