ContactUsLinkComponent
BBL™ Dermatophyte Test Medium Base, BD Biosciences
BBL™ Dermatophyte Test Medium Base, BD Biosciences
Catalog # CA90000-606
Supplier:  BD
CAS Number:  
BBL™ Dermatophyte Test Medium Base, BD Biosciences
Catalog # CA90000-606
Supplier:  BD
Supplier Number:  212330
CAS Number:  
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.
To order chemicals, medical devices, or other restricted products, please provide identification that includes your business name and shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of identification are:
  • • Issued document with your organization's Federal Tax ID Number
  • • Government issued document with your organization's Resale Tax ID Number
  • • Any other Government ID that includes the business name and address
Avantor will not lift restrictions for residential shipping addresses.
Specifications
  • Description:
    BBL™ Dermatophyte Test Medium Base
  • Size:
    500 g
  • Environmentally Preferable:
  • Cat. no.:
    CA90000-606
  • Supplier No.:
    212330
Specifications
About this item
Dermatophyte Test Medium Base is a specialized agar used as a highly selective medium for the isolation of dermatophytes.
  • Highly selective medium
  • Helps in the isolation and early recognition
  • For members of the Microsporum species and Trichophyton species
Nondermatophytes can be recognized by the absence of color change. Rapidly growing species may effect a complete color change within three days, while slow growers will change color in a longer time.