Workrite® FR Nomex® Women's Knit Cuff Flame Resistant Lab Coat with Westex® ShieldCXP™ Chemical Splash Protection, Workrite Uniform Co
Supplier: Bulwark Protection
Certificates
About this item
Women's Lab Coats feature lightweight FR Nomex® IIIA fabric with Westex® ShieldCXP™ Chemical Splash Protection finish
- Black lay-down collar for easy recognition as a Chemical Splash Protection lab coat
- Nomex® knit cuffs (FR only) typically covered by gloves when working with chemicals
- Waistband in back for tailored fit
- Concealed snap front closure
- Splash resistant side pass-through for easy inner pocket access
- One double-stitched chest pocket and two large double-stitched lower front pockets with snap flaps
The fabric resists penetration and wicking of any inadvertent liquid chemical splash.
Certifications: UL certified to NFPA 2112.
Specifications
- Catalog No:
- 75795-946
- 75795-966
- 75795-944
- 75795-950
- 75795-970
- 75795-948
- 75795-968
- 75795-954
- 75795-952
- Garment application:
- Laboratory
- Laboratory
- Laboratory
- Laboratory
- Laboratory
- Laboratory
- Laboratory
- Laboratory
- Laboratory
- Risk area:
- Flame-retardant
- Flame-retardant
- Flame-retardant
- Flame-retardant
- Flame-retardant
- Flame-retardant
- Flame-retardant
- Flame-retardant
- Flame-retardant
- Gender:
- For ladies
- For ladies
- For ladies
- For ladies
- For ladies
- For ladies
- For ladies
- For ladies
- For ladies
- Sleeve or leg length:
- Long
- Long
- Long
- Long
- Long
- Long
- Long
- Long
- Long
- Closure type:
- Press studs / snaps
- Press studs / snaps
- Press studs / snaps
- Press studs / snaps
- Press studs / snaps
- Press studs / snaps
- Press studs / snaps
- Press studs / snaps
- Press studs / snaps
- Fabric:
- Nomex® IIIA fabric
- Nomex® IIIA fabric
- Nomex® IIIA fabric
- Nomex® IIIA fabric
- Nomex® IIIA fabric
- Nomex® IIIA fabric
- Nomex® IIIA fabric
- Nomex® IIIA fabric
- Nomex® IIIA fabric