Viva C4 HPLC Columns, Restek
Supplier: Restek
Certificates
About this item
Viva columns are based on a wide pore material we designed for optimal large-molecule separations.
- USP L26
- Excellent for separating peptides or proteins
- Rugged, spherical particles with 300 Å pore size
- High proportion of pore/surface area available to large molecules
In developing Viva silica, we found that although many commercial wide-pore silicas meet the standard 300 Å mean pore size, most have very broad distributions about this mean, with a significant portion of their pore volume falling below 150 Å.
Base-deactivated, wide-pore packing exhibits excellent peak shape for a wide range of compounds. Less retention in reversed-phase assays than Viva C18 or Viva C8.
This means a large portion of the surface area is unavailable to larger molecules. Viva columns have a narrow distribution around the mean pore size, permitting a larger portion of the silica surface to play a role in the separation.
Specifications
- Catalog No:
- 10803-446
- 10058-132
- Description:
- Viva HPLC Column
- Viva HPLC Column
- Carbon load:
- Carbon Load: 3.5%
- Carbon Load: 3.5%
- Endcapped:
- Yes
- Yes
- pH range:
- 2.5 to 8
- 2.5 to 8
- Temperature limit:
- 80°C
- 80°C
Specifications
Customers Who Bought This Also Bought