Microfiltration Assembly with Fritted Glass Support and Ground Joint, Kimble Chase
Supplier: DWK Life Sciences (KIMBLE)
Certificates
About this item
This 90 mm filtration assembly with fritted glass support is designed for large volume filtration analysis of corrosive liquids or samples with high particulate loads that would tend to clog a 47 mm diameter filter.
- This unit is also recommended for filtering large volumes of HPLC solvents
- Manufactured from 33 expansion, low extractable borosilicate glass conforming to USP Type I and ASTM E438, Type I, Class A requirements
Customers Who Bought This Also Bought