Aayan® Economy Suture Training Kit
Catalog #: 470222-554
Supplier: DR INSTRUMENTS INC. SE
Specifications
- Description:Basic Suture Training Set
- No. of Lab Groups:1
- Cat. no.:470222-554
- Supplier No.:10SK_B
Specifications
About this item
The Aayan™ Economy Suture Training Kit is ideal for medical and veterinary students learning surgical techniques, as well as professional surgeons looking to improve their skills.
- Suture Pad Can Be Used In Or Out Of The Case
- Convenient Storage And Carrying Case
This kit provides all the necessary tools for students to enhance their technique safely and practically. This 8 piece kit comes neatly packed away in a zippered case for easy access and storage.
Contents:
Zippered Case
Needle Holder
Forceps
Stitching Scissors
Disposable Scalpel
Sutures
Skin Pad
Caution: Sutures are intended for Training use only, and are not intended for human or animal use.
Customers Who Bought This Also Bought