ContactUsLinkComponent
Vancomycin Disks, Hardy Diagnostics
Vancomycin Disks, Hardy Diagnostics
Catalog #: 75813-974
Supplier:  Hardy Diagnostics
Product available on GSA Advantage®
CAS Number:  
Vancomycin Disks, Hardy Diagnostics
Catalog #: 75813-974
Supplier:  Hardy Diagnostics
Product available on GSA Advantage®
CAS Number:  
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.
To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
  • • State issued document with your organization's Federal Tax ID Number
  • • State issued document with your organization's Resale Tax ID Number
  • • City or County issued Business License
  • • State Department of Health Services License
  • • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Specifications
  • Description:
    Vancomycin Disks
  • Size:
    50 disks
  • Cat. no.:
    75813-974
  • Supplier No.:
    Z7501
Specifications
About this item
Vancomycin disks are used for the differentiation of anaerobic bacteria based on susceptibility to vancomycin.
  • Differentiation of anaerobic bacteria
  • Spring loaded for smooth and reliable dispensing
For the differentiation of anaerobic bacteria based on susceptibility to vancomycin. Each cartridge contains 50 disks that are spring loaded for smooth and reliable dispensing from the single metal disk dispenser.
Customers Who Bought This Also Bought