Regan-Lowe Semi Solid Agar Deep, Hardy Diagnostics
Catalog #: 75814-678
Supplier: Hardy Diagnostics
Product available on GSA Advantage®
CAS Number:
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.
To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Specifications
- Format:13×100 mm tube
- Description:Regan-Lowe Semi Solid Agar Deep
- Size:Pack of 20
- Volume:4 ml
- Supplier No.:Q32
Specifications
About this item
Regan-Lowe Semi Solid Agar Deep is used for the selective isolation of Bordetella pertussis.
- Optically clear, shatter resistant tube
- Selective Bordetella pertussis isolation
Charcoal Blood Agar with 10% Horse Blood and Cephalexin, for the selective isolation of Bordetella pertussis.
Customers Who Bought This Also Bought