To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Specifications
- Format:16×125 mm tube
- Description:Lactobacilli MRS Broth
- Size:Pack of 20
- Minimum Order Quantity:5
- Volume:9 ml
- Supplier No.:K17
Specifications
About this item
Lactobacilli MRS Broth is used for the cultivation of lactobacilli and differentiation of gram-positive organisms.
- Contains a durham tube
- Cultivation of lactobacilli
- Differentiation of gram-positive organisms
For the cultivation of lactobacilli and differentiation of gram-positive organisms. Contains a durham tube.