Water, deionized sterile
Catalog #: 75814-006
Supplier: Hardy Diagnostics
Product available on GSA Advantage®
CAS Number: 7732-18-5
Specifications
- Size:Pack of 10
- Cat. no.:75814-006
- Supplier No.:U284
Specifications
Specification Test Results
- Water, deionized sterile:
Specification Test Results
About this item
Synonyms: Hydrogen oxide
:Length:
:Width:
:Height:
:Ø
:Thickness:
:Boiling Pt:
:Boiling Pt:
:Melting Pt:
:Melting Pt: 0 °C
:Flash Pt:
:Flash Pt:
:M.W. 18.02 g/mol
:Water
:0
:100
:12,10175
Formula:H₂O
MW:18.02 g/mol
MDL Number:MFCD00011332
CAS Number:7732-18-5
Customers Who Bought This Also Bought