1,4-Dioxane ≥99.9%, HiPerSolv CHROMANORM® for HPLC, VWR Chemicals BDH®
Catalog #: BDH83628.400
Supplier: VWR International
CAS Number: 123-91-1
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.
To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Specifications
- Packaging:Glass bottle, amber, poly-coated
- Size:4 L
Specifications
Specification Test Results
- Assay (by GC-FID):Min 99.9%
- Color:Max 10 APHA
- Peroxides/Epoxides (as Hydrogen peroxide at time of packaging):Max 0.5%
- Residue after evaporation:Max 5 ppm
- Water (by KF coulometry):Max 0.5%
- Transmittance at 215 nm:Min 10 T(%)
- Transmittance at 225 nm:Min 25 T(%)
- Transmittance at 250 nm:Min 55 T(%)
- Transmittance at 295 nm:Min 98 T(%)
- Transmittance at 400 nm:Min 98 T(%)
- Optical absorbance at 215 nm:Max 1.00 AU
- Optical absorbance at 225 nm:Max 0.60 AU
- Optical absorbance at 250 nm:Max 0.25 AU
- Optical absorbance at 295 nm:Max 0.01 AU
- Optical absorbance at 400 nm:Max 0.01 AU
- 1,4-Dioxane ≥99.9%, HiPerSolv CHROMANORM® for HPLC, VWR Chemicals BDH®:
Specification Test Results
About this item
Synonyms: Glycolethylether,1,4-Dioxacyclohexane,Dioxane,p-Dioxane,1
Boiling Pt:101.1 °C (1013 hPa)
Melting Pt:11.8 °C
ADR:3,II
:Length:
:Width:
:Height:
:Ø
:Thickness:
:99,9
Flash Pt:12 °C
:Boiling Pt:
:Boiling Pt: 101.1 °C (1013 hPa)
:Melting Pt:
:Melting Pt: 11.8 °C
:Flash Pt:
:Flash Pt: 12 °C
:M.W. 88.11 g/mol
:1,4-Dioxane
:11.8
:101.1
MW:88.11 g/mol
MDL Number:MFCD00006571
UN:1165
:3
:II
:12
Formula:C₄H₈O₂
CAS Number:123-91-1
Customers Who Bought This Also Bought