To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Specifications
- Size:30 mg
- Enzyme name:Proteinase K
- Enzyme activity:20 U/mg
- Concentration:20 mg/ml
- Cat. no.:CA10755-738
- Supplier No.:845-CH-0010030
Specifications
About this item
innuPREP proteinase K is a highly active recombinant protease from Pichia pastoris with endo-and exoproteolytic activity.
- Contains no RNases or DNases and virtually no DNA
- Consistent quality and performance
- Stable over a broad pH range from 4 to 12.5
- Ideal for preparing cell lysates for subsequent nucleic acid isolation
This enzyme is extraordinarily effective against native proteins and can be used for quickly inactivating endogenous RNases and DNases. Proteinase K is suitable for isolating nucleic acids for use in amplification reactions, for isolating native RNA and DNA form tissues and cell lines, for promoting cell lysis by activating a bacterial autolysis factor and for modifying proteins and/or glycoproteins on cell surfaces (for membrane structure analyses).