ULTRA-WARE® THF-Resistant Five Valve Recirculation/Filtration Cap, Kimble Chase, DWK Life Sciences
Catalog #: KT953983-6347
Supplier: DWK Life Sciences (KIMBLE)
Specifications
- Cap Size:GL 45
- Ø:47 mm
- Description:THF-Resistant Five Valve Recirculation/Filtration Cap
- Cat. no.:KT953983-6347
- Supplier No.:953983-6347
Specifications
About this item
THF-Resistant filtration cap ideal for aggressive organic solutions such as chlorinated hydrocarbons, ethers and ketones.
- Cap body is manufactured from glass-filled PTFE
- Cap will fit any reservoir or bottle with a GL 45 thread
THF-resistant version has 316 stainless steel which prevents the attack of CTFE valve stems and TFE/propylene o-rings byTetrahydrofluran (THF).
Customers Who Bought This Also Bought