To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Specifications
- Antibody type:Primary
- Antigen name:Actin, Alpha, Cardiac Muscle 1
- Clonality:Monoclonal
- Clone:mAbGEa
- Gene ID:70
- Host:Mouse
- Isotype:IgG1
- Reactivity:Dog,Rat,Mouse,Zebrafish,Yeast,Hamster,Drosophila,Plant,Pig,Bacteria,Sheep,Human,Bovine,Xenopus laevis
- Antigen symbol:ACTC1
- Conjugation:Unconjugated
- ImmunoChemistry:Yes
- ImmunoFluorescence:Yes
- Size:100 μl
- Western blot:Yes
- Antigen synonyms:CMD1R|LVNC4|CMH11|ACTC|ASD5
- Storage buffer:Ascites containing 0.05 % sodium azide
- Storage temperature:–20 °C, avoid freeze/thaw cycles
- Immunogen:Purified Arabidopsis actin protein.
- Cat. no.:10168-838
- Supplier No.:GTX80809
Specifications
About this item
Mouse monoclonal antibody [mAbGEa] to Actin
Type: Primary
Antigen: ACTC1
Clonality: Monoclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human, Mouse, Dog, Drosophila, Pig, Rat, Zebra Fish, Yeast, Hamster, Sheep, Xenopus laevis, Bacteria, Plant, Bovine