Anti-SLC25A10 Rabbit Polyclonal Antibody
Catalog #: 10164-862
Supplier: Genetex
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.
To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Specifications
- Antibody type:Primary
- Antigen name:Solute Carrier Family 25 (Mitochondrial Carrier; Dicarboxylate Transporter), Member 10
- Clonality:Polyclonal
- Gene ID:1468
- Host:Rabbit
- Isotype:IgG
- Reactivity:Human
- Antigen symbol:SLC25A10
- Conjugation:Unconjugated
- ImmunoChemistry:Yes
- Size:100 μg
- Antigen synonyms:DIC
- Storage buffer:Aqueous buffered solution containing 100ug/ml BSA, 50% glycerol and less than 0.09% sodium azide.
- Storage temperature:Keep as concentrated solution. Aliquot and store at –20 °C or below. Avoid multiple freeze-thaw cycles.
- Concentration:1 mg/ml
- Immunogen:KLH conjugated synthetic peptide derived from human SLC25A10
- Purification:Purified by Protein A and peptide affinity chromatography.
- Cat. no.:10164-862
- Supplier No.:GTX51583
Specifications
About this item
Rabbit Polyclonal antibody to SLC25A10
Type: Primary
Antigen: SLC25A10
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Customers Who Bought This Also Bought