Anti-GLS2 Rabbit Polyclonal Antibody
Catalog #: 10164-092
Supplier: Genetex
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.
To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Specifications
- Antibody type:Primary
- Antigen name:Glutaminase 2 (liver mitochondrial)
- Clonality:Polyclonal
- Host:Rabbit
- Isotype:IgG
- Reactivity:Human,Mouse
- Antigen symbol:GLS2
- Conjugation:Unconjugated
- ImmunoChemistry:Yes
- Size:400 μl
- Western blot:Yes
- Antigen synonyms:GLS|GLS2|hLGA|L-glutaminase|LGA|mitochondrial|Glutaminase liver isoform|GA
- Storage buffer:PBS with 0.09% sodium azide
- Storage temperature:Store at –20 °C
- Concentration:0.25 mg/ml
- Immunogen:Synthetic peptide between 505-532 amino acids from the C-terminal region of human GLS2.
- Purification:Affinity purified by Protein A.
- Cat. no.:10164-092
- Supplier No.:GTX50889
Specifications
About this item
Rabbit polyclonal to GLS2
Type: Primary
Antigen: GLS2
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse
Customers Who Bought This Also Bought