Anti-Pan-Ras Rabbit Polyclonal Antibody
Catalog #: 10162-766
Supplier: Genetex
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.
To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Specifications
- Antibody type:Primary
- Antigen name:Pan Ran Asp12
- Clonality:Polyclonal
- Host:Rabbit
- Isotype:IgG
- Reactivity:Human,Rat,Mouse
- Antigen symbol:Pan-Ras
- Conjugation:Unconjugated
- Size:50 μg
- Storage buffer:20 mM ammonium bicarbonate, 100 µg BSA
- Storage temperature:Keep as concentrated solution, aliquot and store at 4°C. Do not freeze.
- Immunogen:Synthetic peptide (KLVVVGADGVGK) corresponding to amino acids 5-16 of Ras (H, K, or N) with Asp at position 12
- Purification:Purified IgG
- Cat. no.:10162-766
- Supplier No.:GTX20064
Specifications
About this item
Rabbit polyclonal to Pan-Ras Asp12
Type: Primary
Antigen: Pan-Ras
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Customers Who Bought This Also Bought