Anti-Ptprc Rat Monoclonal Antibody [clone: RA3.3A1/6.1 (TIB-146)]
Catalog #: 10162-534
Supplier: Genetex
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.
To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Specifications
- Antibody type:Primary
- Antigen name:Protein Tyrosine Phosphatase Receptor Type C
- Clonality:Monoclonal
- Clone:RA3.3A1/6.1 (TIB-146)
- Gene ID:19264
- Host:Rat
- Isotype:IgM
- Reactivity:Mouse
- Antigen symbol:PTPRC
- Conjugation:Unconjugated
- Size:500 μg
- Antigen synonyms:CD45R|B220|CD45|GP180|T200|L-CA|LY5|receptor type C|LCA|protein tyrosine phosphatase
- Storage buffer:PBS, pH 7
- Storage temperature:Keep as concentrated solution. Aliquot and store at –20 °C or below. Avoid multiple freeze-thaw cycles.
- Concentration:1 mg/ml
- Purification:Protein G. The Purity is >95% as determined by SDS-PAGE.
- Cat. no.:10162-534
- Supplier No.:GTX14090
Specifications
About this item
Rat monoclonal antibody [RA3.3A1/6.1 (TIB-146)] to CD45R
Type: Primary
Antigen: Ptprc
Clonality: Monoclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rat
Isotype: IgM
Reactivity: Mouse
Customers Who Bought This Also Bought