Anti-ANKRD26 Rabbit Polyclonal Antibody
Catalog #: 10166-786
Supplier: Genetex
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.
To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Specifications
- Antibody type:Primary
- Antigen name:Ankyrin Repeat Domain 26
- Clonality:Polyclonal
- Gene ID:22852
- Host:Rabbit
- Isotype:IgG
- Reactivity:Human
- Antigen symbol:ANKRD26
- Conjugation:Unconjugated
- Size:100 μl
- Western blot:Yes
- Antigen synonyms:bA145E8.1|THC2
- Storage buffer:1XPBS, 20% Glycerol (pH7). 0.01% Thimerosal was added as a preservative.
- Storage temperature:Keep as concentrated solution. Aliquot and store at –20 °C or below. Avoid multiple freeze-thaw cycles.
- Concentration:1 mg/ml
- Immunogen:Recombinant protein encompassing a sequence within the N-terminus region of human ANKRD26. The exact sequence is proprietary.
- Purification:Purified by antigen-affinity chromatography.
- Cat. no.:10166-786
- Supplier No.:GTX128255
Specifications
About this item
Rabbit Polyclonal antibody to ANKRD26 (ankyrin repeat domain 26)
Type: Primary
Antigen: ANKRD26
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Customers Who Bought This Also Bought