To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Specifications
- Antibody type:Primary
- Antigen name:Transcription Factor 7 (T-Cell Specific, Hmg-Box)
- Clonality:Polyclonal
- Gene ID:6932
- Host:Rabbit
- Isotype:IgG
- Reactivity:Human
- Antigen symbol:TCF7
- Conjugation:Unconjugated
- ImmunoPrecipitation:Yes
- Size:100 μl
- Western blot:Yes
- Antigen synonyms:TCF-1
- Storage buffer:1XPBS, 20% Glycerol (pH7). 0.01% Thimerosal was added as a preservative.
- Storage temperature:Keep as concentrated solution. Aliquot and store at –20 °C or below. Avoid multiple freeze-thaw cycles.
- Concentration:1 mg/ml
- Immunogen:Recombinant protein encompassing a sequence within the center region of human TCF7. The exact sequence is proprietary.
- Purification:Purified by antigen-affinity chromatography.
- Cat. no.:10166-606
- Supplier No.:GTX113851
Specifications
About this item
Rabbit Polyclonal antibody to TCF7 (transcription factor 7 (T-cell specific, HMG-box))
Type: Primary
Antigen: TCF7
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human