Sediment Settler Kit
Catalog #: 470039-542
Supplier: Avantor
Specifications
- Description:Sediment Settler Kit
- No. of Lab Groups:15
- Cat. no.:470039-542
- Supplier No.:WARD470039-542
Specifications
About this item
Discover What Sediment is and its Impact on the Environment
- Materials for 30 students
- Grades 4-8
In this hands-on project, students assemble their own Sediment Settler by adding four types of particles, in varying sizes and densities, to a plastic tube with screw cap. Once assembled, students observe their Sediment Settler before and after adding water, and compare settling time with particle size.
Includes instructions for addressing two levels of National Standards.
Tube size: 15 x 2.5 cm
Customers Who Bought This Also Bought