Absorb-&-Lock® Bio-Fluids Absorbent, PIG®
Catalog #: 10861-014
Supplier: New Pig
Specifications
- Description:PIG® Absorb-&-Lock® Bio-Fluids Absorbent
- Size:4 containers per box
- Cat. no.:10861-014
- Absorbency:8 gal./container
- Supplier No.:PLP503
Specifications
About this item
Permanently solidifies to lock in absorbed liquids so they won't leak or liquify when the absorbents are handled or placed under pressure.
- Contains chlorine to kill pathogens
- Clean scent helps reduce offensive odors
- Environmentally-friendly product
Contains chlorine to kill pathogens, reduce odors and help you meet OSHA standards. Clean scent helps reduce offensive odors during and after cleanup. Environmentally-friendly product absorbs biological fluids including blood, urine and vomit.
Customers Who Bought This Also Bought