Ashcroft® Test Gauges, Restek
Supplier: Restek
Certificates
About this item
High-accuracy test gauges are recommended for verifying the vacuum/pressure in canisters before and after sampling.
- Accurate measurement of vacuum to –30" Hg and pressure to 60 psi
- Available in both analog and digital formats
- Accuracy to +/– 0.25%
- Gauge connector to canister valve available
The 6-inch face on the analog gauge allows for easy reading. The digital gauge operates on two AAA batteries and offers an unambiguous readout. Both gauges have an accuracy of +/– 0.25% and all-metal wetted parts.
Customers Who Bought This Also Bought