Spectra Gas 7621 High-Purity VOC Regulators, Restek
Catalog #: 10862-704
Supplier: Restek
Specifications
- Description:Regulator, Spectra Mini SS High Purity 1 Stage Regulator for 6R Cylinder - CGA 180
- Cat. no.:10862-704
- Supply Pressure Gauge:0 - 30 psig
- Supplier No.:21572
Specifications
About this item
Spectra Gas 7621 High-Purity VOC Regulators with two pressure gauges.
- Single-stage, stainless steel
- Two pressure gauges and CGA-180 fitting
- Stainless steel diaphragm and Kel-F® seat
- Accurate pressure control even at low flow rates
Customers Who Bought This Also Bought