Thermal Desorption Unit Tubes (Calibration), Restek
Supplier: Restek
Certificates
About this item
High-quality thermal desorption tubes are suitable for ppt to ppm concentrations of volatile organic compounds (VOCs) in ambient, indoor, personal, and industrial hygiene environments.
- Use corrosion-resistant glass tubes for excellent inertness
- Allow monitoring of sorbent bed condition
- Choose stainless steel tubes for greater durability in the field
- No sampling pump necessary for passive sampling with diffusion caps
- Individually etched with unique serial number for convenient sample identification
- The most reliable barcode "code 128" format on tube for recording and tracking
- Each tube has an arrow indicating flow direction to reduce errors during use
Fit Markes (ULTRA and UNITY™), PerkinElmer, and Shimadzu thermal desorbers. Packed tubes come with a report detailing the total mass of sorbent in the tube; conditioned tubes also include a blank chromatogram.
Certifications: US EPA TO-17; ASTM D6196; NIOSH 2549; DIN EN ISO 16017
Customers Who Bought This Also Bought