Axygen® Horizontal Gel Electrophoresis Units, Corning
Supplier: Corning
Certificates
About this item
Axygen® horizontal gel box is a high quality, full-featured unit designed for exceptional durability and efficiency. These gel boxes feature leak-free external gel casting, which allows the user to cast one gel outside of the box while another gel is running inside. Boxes from 7, 10, 15 and 20cm
- Flexibility – Each unit comes with 2 sizes of gel trays to optimize applications
- External gel casting to gel box allows for running gels while casting the next one
- Ease of use – the Axygen contrast enhancer makes loading gels easier
- Rugged injection molding is supported by a 3-year warranty
- Larger units come with recirculation ports allowing for buffer exchange
Certifications: CE marked.
Specifications
- Catalog No:
- 75799-872
- 75800-104
- 75801-458
- 75800-102
- Model:
- HGB-10
- Tank volume:
- 300 ml (buffer volume)
- 1500 ml (buffer volume)
- 225 ml (buffer volume)
- 500 ml (buffer volume)
- Width:
- 12.5 cm
- 23 cm
- 9 cm
- 17.5 cm
- Length:
- 22 cm
- 39.5 cm
- 21 cm
- 26.5 cm
Specifications
Related Information
Customers Who Bought This Also Bought