To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Specifications
- Packaging:Bottle
- Source:Plasma
- Species:Bovine
- CAS No.:9002-04-4
- Size:25 KU
- Storage conditions:−10 °C
- Form:Off-white solid
- Enzyme name:Thrombin
- Enzyme activity:90 - 300 u/mg protein
- pH:7.0 - 8.0
- Supplier No.:0219990725
Specifications
About this item
Thrombin is the final coagulation protease in regard to hemostasis, promoting both procoagulant and anticoagulant effects. It is a lyophilized powder containing sucrose, sodium chloride and Tris. The predominant form of thrombin in vivo is the zymogen, prothrombin (factor II), which is produced in the liver. The concentration of prothrombin in normal human plasma is 5–10 mg/dL. Prothrombin is a glycoprotein with a glycan content of ~12%.
- Enzyme Commission Number: E.C.3.4.21.5
- Extinction coeficient: E2801% = 18.3 (human); E2801% = 19.5 (bovine)
12%.
Serine protease that selectively cleaves Arg-Gly bonds in fibrinogen to form fibrin and fibrinopeptides A and B.
Thrombin is used throughout the diagnostics industry in a variety of coagulation assays, clotting factor tests and defibrination of blood or plasma for serum controls. It is also used for site specific cleavage of recombinant fusion proteins, and in biochemical and medical research applications. It is manufactured from New Zealand-sourced bovine plasma using prothrombin activated with thromboplastin extracted from bovine lung tissue. It is lyophilized in glass vials under vacuum to optimize product performance. Low specific activity thrombin has 90 - 500 NIH units/mg and greater than 50% total protein. It has been used in a study to assess persistent hypocoagulability in patients with septic shock.