To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Specifications
- Packaging:Bottle
- Source:Serum
- Species:Bovine
- CAS No.:9048-46-8
- Size:50g
- Bioburden:<100 cfu/g
- Storage conditions:+4 °C
- Endotoxin content:<1.0 EU/mg
- Form:Powder
- Protein synonyms:BSA|Fraction V|Cohn Fraction V
- Protein/peptide name:Bovine Serum Albumin
- Purity:>97% Total Protein
- Solubility:Dissolves completely in 20 min at 20-25 °C (20%)
- Molecular weight:66 kDa
- Endotoxin level:low
- Colour:Off-white to Light Yellowish to Brown
- Concentration:Total Protein: ≥95%
- pH:6.5 - 7.5 (4% aq soln)
- Grade:Stem Cell Grade
- Cat. no.:10273-262
- Supplier No.:0219989650
Specifications
About this item
Bovine albumin is a single polypeptide chain consisting of approximately 583 to 595 amino acid residues and no carbohydrates. At pH 5-7 it contains 17 intrachain disulfide bridges and 1 sulfhydryl group.
- Stem Cell Grade
- Presentation: Off-white to Light Yellowish to Brown Powder
- Isoelectric point (pl): Endogenous material- 4.7, 4.9; Fatty acid depleted- 5.3(In Water at 25 °C)
- Moisture: ≥5%
Albumin binds water, Ca2+, Na+, and K+. Due to a hydrophobic cleft, albumin binds fatty acids, bilirubin, hormones and drugs. The main biological function of albumin is to regulate the colloidal osmotic pressure of blood and to a lesser degree to provide cellular nutrition. Human and bovine albumins contain 16% nitrogen and are often used as standards in protein calibration studies. Albumin is used to solubilize lipids, stabilize protein solutions, and is also used as a blocking agent in Western blots or ELISA applications. Globulin free albumins are suitable for use in applications where no other proteins should be present such as electrophoresis.
Albumins are a group of acidic proteins which occur in the body fluids and tissues of mammals and in some plant seeds. Serum and plasma albumin is carbohydrate-free and comprises 55-62% of the protein present. However, only about 40% of the total albumin in the body is in the circulating plasma at one time with the remainder being in extracellular spaces with which there is, in general, equilibration about every 24 hours.
Albumin loses its helical structure rapidly in 2-4 M urea and 4 M guanidinium chloride. The unfolding is reversible, even, to some extent, after concomitant reduction of all disulfide bonds. Reduced albumin can reform into structures capable of binding antibodies and other ligands. Formation of SS bonds is rapid but the return of native structure is slow and some wrong disulfide pairings may persist. After oxidation of the cystine bridges, however, native structure is irreversibly lost.
Albumins are readily soluble in water and can only be precipitated by high concentrations of neutral salts such as ammonium sulfate. The solution stability of albumin is very good (especially if the solutions are stored aliquoted and frozen). Albumin is readily coagulated by heat. When heated to 50 °C or above, albumin quite rapidly forms hydrophobic aggregates which do not revert to monomers upon cooling. Dissolves completely in 20 min at 20-25 °C (20%).