Aluminium Sample Pans, PerkinElmer
Supplier: PERKINELMER SCIENTIFIC CANADA ULC
Certificates
About this item
Each sample pan and cover has an operating range of -170° to 600 °C, and an ambient max pressure.
- Can be used with nonvolatile solid samples
- Crimped design
- Not hermetically sealed
These sample pans can be used with nonvolatile solid samples, such as plastics and polymers, and are routinely used for applications such as polymer melting, crystallization, glass transitions of polymers, thermoplastics, and thermosets. Pans are crimped, but not hermetically sealed. Not for use with Diamond DSC, PYRIS 1 DSC, or DSC 7 Autosamplers.
Customers Who Bought This Also Bought