Prime/Purge Valve Repair Kit for Prime/Purge Valve, Restek
Catalog #: 10853-752
Supplier: Restek
Specifications
- Description:Prime/Purge Valve Repair Kit for Prime/Purge Valve
- Cat. no.:10853-752
- Includes:Soft seal (2), PTFE O-ring (2), 7/32 hex key
- Supplier No.:26407