Plunger Seals for Thermo HPLC Systems, Restek
Supplier: Avantor
Certificates
About this item
Graphite-filled PTFE; best for organic solvents
Ultra-high molecular weight polyethylene (UHMWPE); increased resistance to abrasion, best for buffers.
Customers Who Bought This Also Bought