Plunger Seal, Superseal for Thermo HPLC Systems, Restek
Catalog #: 10853-332
Supplier: Restek
Specifications
- Description:Plunger Seal, Gold Superseal for SP8800 and P-Series Pumps, Similar to SP #A2962-010
- Model:8800, 8810, ISOCHROM, P1000, P2000, P4000
- Cat. no.:10853-332
- Supplier No.:25478
Specifications
About this item
Ultra-high molecular weight polyethylene (UHMWPE); increased resistance to abrasion, best for buffers.
Customers Who Bought This Also Bought