Sapphire Plunger Assembly Kit for Waters HPLC Systems, Restek
Catalog #: 10853-568
Supplier: Restek
Specifications
- Description:Sapphire Plunger Assembly Kit for Waters 515, Similar to Waters #WAS207069
- Model:515, 1515, 1525
- Includes:Sapphire plunger, pivot insert, pivot guide, washer, spring, retaining ring
- Supplier No.:25384