Plunger Assembly for Shimadzu HPLC Systems, Restek
Supplier: Restek
Certificates
Customers Who Bought This Also Bought
Sulfuric acid 1.0 N in aqueous solution, VWR Chemicals BDH®
Supplier: VWR International
$64.40 to $734.93
Glycerine ≥99.7%, Laboratory Reagent, VWR Chemicals BDH®
Supplier: VWR International
$881.80 to $2,807.56
Methanol ≥99.9%, OmniSolv®, gradient grade for HPLC, Supelco®
Supplier: MilliporeSigma
$635.20 to $2,541.50
NAB Nanosep® Centrifugal Devices for RNA and DNA Extraction, Cytiva (Formerly Pall Lab)
Supplier: Cytiva
$141.42