Low Pressure Seal for PerkinElmer HPLC Systems, Restek
Catalog #: 10853-730
Supplier: Restek
Specifications
- Description:Low Pressure Seal
- Model:Int. 4000, 1, 10, 2, 200 Series, 250, 3, 3B, 4, 400, 410, 620
- Cat. no.:10853-730
- For Use With:PE Series 200 pumps
- Supplier No.:25435
Specifications
About this item
These seals made of ultra-high molecular weight polyethylene (UHMWPE); increased resistance to abrasion, best for buffers.
Customers Who Bought This Also Bought