Standard High Pressure Seal for PerkinElmer HPLC Systems, Restek
Catalog #: 10853-508
Supplier: Restek
Specifications
- Description:Standard High Pressure Seal
- Cat. no.:10853-508
- For Use With:PE Series 200 pumps 1,2,3,3B,4,10,250,400,410,620, Int. 4000 Pump
- Supplier No.:25434
Specifications
About this item
Ultra-high molecular weight polyethylene (UHMWPE); increased resistance to abrasion, best for buffers.
Customers Who Bought This Also Bought