High Pressure Seal Kit for PerkinElmer HPLC Systems, Restek
Catalog #: 10853-588
Supplier: Restek
Specifications
- Description:High Pressure Seal Kit for PE Series 200 pumps 1,2,3,3B,4,10,250,400,410,620, Int. 4000 Pump, Similar to PE# 0254-0275
- Model:Int. 4000, 1, 10, 2, 200 Series, 250, 3, 3B, 4, 400, 410, 620
- Cat. no.:10853-588
- Includes:Pump seals (4), backup rings (4), O-rings (4)
- Supplier No.:25433