FlatLine Pulse Damper, Restek
Catalog #: 10853-638
Supplier: Restek
Specifications
- Description:FlatLine Pulse Damper
- Cat. no.:10853-638
- Supplier No.:25340
Specifications
About this item
The ASI FlatLine Pulse Damper combines performance and reliability in a simple, easy-to-use housing.
- Rupture-proof, no diaphragm—minimal risk of failure or leaks
- Clean flush-out design—no sample carryover
- Low internal volume—negligible effect on analyte bandwidth
Standard 10-32 inlet and outlet ports allow quick connection into virtually any HPLC system. Solid core technology provides reliable long-term operation without the downtime associated with ruptured or leaking membrane dampers. 600–700 µL internal volume at ambient pressure. Maximum pressure: 6,000 psi (41,369 kpa)
Customers Who Bought This Also Bought