ValvTool Wrench, Restek
Catalog #: 10853-760
Supplier: Restek
Specifications
- Description:ValvTool Wrench for 1/4" Nuts
- Cat. no.:10853-760
- Supplier No.:25321
Specifications
About this item
The ValvTool is a time-saving device that provides easy access to many hard-to-reach Rheodyne® or Valco® valves.
For 1/4" nuts.
Customers Who Bought This Also Bought