Low-Pressure CPI Inlet Filter for Mobile Phase Reservoir, Restek
Catalog #: RK25009
Supplier: Restek
Specifications
- Description:Low-Pressure Inlet Filter, 1/8" 10 µm CPI
- Cat. no.:RK25009
- Supplier No.:25009
Specifications
About this item
A 316 stainless steel knurled cap and Tefzel® CPI ferrule seal to ¹/₈" Ext.Ø PTFE tubing when finger-tightened onto the precision-machined filter holder.
The filter element is replaceable. Standard 10 µm porosity protects delicate pump components from particles but introduces very little pressure drop. ¹/₈" Ext.Ø. May be used as a helium sparging diffuser.
Customers Who Bought This Also Bought