To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Specifications
- Description:Gifu Anaerobic Broth (GAM)
- Size:500 g
- Storage temperature:2 - 8 °C
- Cat. no.:CA11007-214
- Supplier No.:M1801-500G
Specifications
About this item
Gifu Anaerobic Medium (GAM Broth) is recommended as a general culture medium for cultivation and isolation of anaerobic bacteria and to test their susceptibility to antibiotics other than sulpha drugs.
- Light yellow to brownish yellow homogeneous free flowing powder
- Forms amber coloured clear solution in tube
- Reaction of 5.9% w/v aqueous solution at 25 °C, pH: 7.3±0.1
Composition of the medium is as follows: Peptic digest of animal tissue, papaic digest of soyabean meal, protease peptone, digested serum, yeast extract, beef extract, liver extract, dextrose, potassium dihydrogen phosphate, sodium chloride, starch, L-cysteine hydrochloride, sodium thioglycollate.
GAM broth is to be storted below 30 °C and the prepared medium at 2 - 8 °C.