VWR® ULTREX™ Acids Bottle Top Dispenser
Catalog #: CAJT6910-1
Supplier: AVANTOR PERFORMANCE MATERIAL LLC
MOC: Non-RegulatedCertificates
Specifications
- Description:ULTREX™ bottle top dispenser, corrugated case, 1 each
- Cat. no.:CAJT6910-1
- Supplier No.:6910-01
Specifications
About this item
The ULTREX™ bottle-top dispenser made of Teflon PFA or TFM (modified PTFE) reduces the risk of contamination and maintains purity.
- Specifically designed for use with ULTREX™ II acids
- Eliminates leaching and airborne contamination
- Minimize waste and enhance safe handling
Acid pre-cleaned to maintain <0.1 ppb metal blank levels in routine use. All wet parts are constructed with Teflon to maintain product purity.
Delivery: Each unit comes with a PTFE air filter to reduce risk of airborne contamination.
Customers Who Bought This Also Bought