To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Specifications
- Concentration:1000 µg/ml
- Element:Potassium
- Matrix:5% HNO₃
- Packaging:Narrow mouth poly liquid bottle
- Size:150 ml
- Supplier No.:6464-04
Specifications
About this item
The standard is prepared by the careful dilution of the metal or its salt, usually 99.99% pure as verified by arc emission spectrometry.
- Packaged at a concentration of 1000 µg/ml
- Standardised with NIST standard chemicals
- Maximum shelf life and stability
The final concentration of each lot is determined by titrimetry or gravimetry.