ContactUsLinkComponent
AimStrip® Adulteration 7 Test, Germaine Laboratories
AimStrip® Adulteration 7 Test, Germaine Laboratories
Catalog #: 10024-370
CAS Number:  
Urine Dipstick
AimStrip® Adulteration 7 Test, Germaine Laboratories
Catalog #: 10024-370
CAS Number:  
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.
To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
  • • State issued document with your organization's Federal Tax ID Number
  • • State issued document with your organization's Resale Tax ID Number
  • • City or County issued Business License
  • • State Department of Health Services License
  • • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Specifications
  • Description:
    AimStrip® Adulteration 7 Test
  • Cat. no.:
    10024-370
  • Supplier No.:
    40007
Specifications
About this item
These urine Dipsticks are designed as a semi-quantitative, color comparison screen for the detection of Creatinine, Nitrite, Glutaraldehyde, pH, Specific Gravity, and Oxidants-Pyridinium Chlorochromate (PCC) in human urine
Results are available within 2 to 5 minutes. Results are determined by comparing the reagent pads on the dipstick to a color chart provided on the bottle. Test includes 25 dipsticks per bottle.
This test provides a preliminary screen only, and is intended for forensic use only.
Customers Who Bought This Also Bought