Nitric acid 65%, EMSURE® Reag. Ph. Eur., ISO for analysis, Supelco®
Catalog #: EM1.00456.9026
Supplier: MilliporeSigma
CAS Number: 7697-37-2
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.
To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Specifications
- Packaging:Barrel PE/metal
- Size:25 L
- Cat. no.:EM1.00456.9026
- Supplier No.:1.00456.9026
Specifications
Specification Test Results
- Assay (Alkalimetric):≥65.0%
- Identity:Passes Test
- Appearance:Passes Test
- Color (Hazen):≤10 Hazen
- Density (d 20/20):1.384 - 1.416
- Chloride (Cl):≤0.2 ppm
- Phosphate (PO₄):≤0.2 ppm
- Sulfate (SO₄):≤0.5 ppm
- Heavy Metals (as Pb):≤0.2 ppm
- Ag (Silver):≤0.010 ppm
- Al (Aluminium):≤0.050 ppm
- As (Arsenic):≤0.010 ppm
- Au (Gold):≤0.050 ppm
- Ba (Barium):≤0.010 ppm
- Be (Beryllium):≤0.010 ppm
- Bi (Bismuth):≤0.020 ppm
- Ca (Calcium):≤0.100 ppm
- Cd (Cadmium):≤0.010 ppm
- Co (Cobalt):≤0.010 ppm
- Cr (Chromium):≤0.020 ppm
- Cu (Copper):≤0.010 ppm
- Fe (Iron):≤0.100 ppm
- Ga (Gallium):≤0.050 ppm
- Ge (Germanium):≤0.020 ppm
- In (Indium):≤0.020 ppm
- K (Potassium):≤0.100 ppm
- Li (Lithium):≤0.010 ppm
- Mg (Magnesium):≤0.050 ppm
- Mn (Manganese):≤0.010 ppm
- Mo (Molybdenum):≤0.010 ppm
- Na (Sodium):≤0.200 ppm
- Ni (Nickel):≤0.020 ppm
- Pb (Lead):≤0.010 ppm
- Pt (Platinum):≤0.100 ppm
- Sr (Strontium):≤0.010 ppm
- Ti (Titanium):≤0.020 ppm
- Tl (Thallium):≤0.020 ppm
- V (Vanadium):≤0.010 ppm
- Zn (Zinc):≤0.020 ppm
- Zr (Zirconium):≤0.020 ppm
- Residue on Ignition (as SO₄):≤3 ppm
- Tested by the Dithizone:Passes Test
Specification Test Results
About this item
:63.01
Boiling Pt:121 °C (1013 hPa)
Melting Pt:–32 °C
Density:1.39 g/cm3 (20 °C)
ADR:8 (5.1),II
:Length:
:Width:
:Height:
:Ø
:Thickness:
:M-Clarity™ quality level = MQ300
:Boiling Pt:
:Boiling Pt: 121 °C (1013 hPa)
:Melting Pt:
:Melting Pt: –32 °C
:Flash Pt:
:Flash Pt:
:Density: 1.51 g/cm³ (20 °C)
:M.W. 63.01 g/mol
:Nitric acid
Storage Temperature:Ambient
EINECS:231-714-2
:121 °C (1013 hPa)
UN:2031
:8 (5.1)
:II
:1.51
:GHS06
:GHS03
:GHS05
:MQ300
:12,06672
Formula:HNO₃
MW:63.01 g/mol
MDL Number:MFCD00011349
CAS Number:7697-37-2
Customers Who Bought This Also Bought