VWR® Signature™ Glass Beads, Silica
Supplier: VWR International
Certificates
About this item
Molecular biology grade silica beads, grinding media for VWR® Signature™ Pulsing Vortex Mixers.
- Treated to be DNase/RNase-free
- Useful for preparing samples for molecular biology applications
Specifications
- Catalog No:
- 12621-154
- 12621-148
- 12621-160
- Environmentally Preferable:
- Waste Reducing
- Waste Reducing
- Waste Reducing
- Environmental certification:
- RoHS
- RoHS
- RoHS