About this item
Premium Grade FBS is widely used by cell culturists requiring high quality, high performance FBS for a wide variety of applications.
- 100% US Origin
- High quality, high performance FBS for a variety of applications
- Preservation of native, growth-promoting factors and minimization of growth-inhibiting factors
- ISIA Certified Supply Chain for Traceability
The care and attention we devote to the manufacture of our products generate final test results that are substantially lower than product specifications
Avantor Seradigm is a supplier of animal sera that provides the cell culture community with access to the most reliable supply of high performance, exceptional quality Fetal Bovine Serum (FBS). Our approach to sourcing and manufacturing results in product performance and quality unlike any other.
Example Certificates of Analysis show typical results.
To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Specifications
- Catalog No:
- 97068-085
- 97068-086
- 97068-088
- 89510-194
- 89510-196
- 97068-091
- Endotoxin:
- ≤5 EU/ml
- ≤5 EU/ml
- ≤5 EU/ml
- ≤5 EU/ml
- ≤5 EU/ml
- ≤5 EU/ml
- Hemoglobin:
- ≤25 mg/dl
- ≤25 mg/dl
- ≤25 mg/dl
- ≤25 mg/dl
- ≤25 mg/dl
- ≤25 mg/dl
- Storage temperature:
- Frozen
- Frozen
- Frozen
- Frozen
- Frozen
- Frozen
Specifications
Related Information
Certificates
Search by Lot NumberAvantor Seradigm Fetal Bovine Sera (FBS) and FBS Alternatives Avantor Seradigm Performance Challenging Fetal Bovine Serum (FBS) and FBS alternatives Avantor Seradigm Premium Grade Fetal Bovine Serum (FBS) Production Insight Solutions for Controlled Environments and Manufacturing Processes Production insight Solutions for Critical Environments and Manufacturing Processes
Product Literature
ghs_1500