Polar ISO Column Test Mix, Restek
Catalog #: 10058-062
Supplier: Restek
CAS Number:
Specifications
- Description:Polar ISO Column Test Mix
- Cat. no.:10058-062
- Supplier No.:35103
Specifications
About this item
Polar ISO column test mix includes aniline, methyl dodecanoate, 2-chlorophenol, n-nonadecane, 2-dodecanol, 2-nonanone, n-heptadecane, 1-octanol.
- 250 μg/mL each in 1,2-dichloroethane, 1 mL/ampoule
Customers Who Bought This Also Bought