PBT Screw Caps for DURAN® Laboratory Glass Bottles, Wheaton, DWK Life Sciences
Supplier: DWK Life Sciences (KIMBLE)
Certificates
About this item
The cap is available with a PTFE coated silicone seal or as a screw connection cap with aperture.
- Autoclavable
- Chemically resistant
- Temperature resistant to 180°C
Related Information
Certificates
Search by Lot NumberDWK DURAN® Innovative Bottle Systems
Product Literature
DWK Life Sciences Return to Research Promo 2021
Promotion Material
Customers Who Bought This Also Bought